ADVH49B82017 AChemBlock

(2R,5R)-2,5-Dimethylmorpholine hydrochloride

  • Cas Number : 1639886-52-4
  • Molecular Weight : 151.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2r-5r-2-5-dimethylmorpholine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=K16121
Availability details for
Quantity
  • Availability
  • Add to Cart