ADVH49B8201A AChemBlock

6-chloro-1,2,3,4-tetrahydroquinoline

  • Cas Number : 49716-18-9
  • Molecular Weight : 167.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-1-2-3-4-tetrahydroquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75365
Availability details for
Quantity
  • Availability
  • Add to Cart