ADVH49B8201C AChemBlock

5-Methyl-4-azaindole

  • Cas Number : 4943-67-3
  • Molecular Weight : 132.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-4-azaindole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72415
Availability details for
Quantity
  • Availability
  • Add to Cart