ADVH49B82023 AChemBlock

5-bromo-2-chloro-4,6-dimethylpyrimidine

  • Cas Number : 4786-72-5
  • Molecular Weight : 221.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-chloro-4-6-dimethylpyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75421
Availability details for
Quantity
  • Availability
  • Add to Cart