ADVH49B82025 AChemBlock

methyl 4-hydroxypyrimidine-5-carboxylate

  • Cas Number : 4774-35-0
  • Molecular Weight : 154.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-hydroxypyrimidine-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71315
Availability details for
Quantity
  • Availability
  • Add to Cart