ADVH49B82027 AChemBlock

3-nitro-1H-indole

  • Cas Number : 4770-03-0
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-nitro-1h-indole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75433
Availability details for
Quantity
  • Availability
  • Add to Cart