ADVH49B82029 AChemBlock

6-nitro-1H-indole

  • Cas Number : 4769-96-4
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-nitro-1h-indole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71317
Availability details for
Quantity
  • Availability
  • Add to Cart