ADVH49B8202C AChemBlock

2-fluoropropanedioic acid

  • Cas Number : 473-87-0
  • Molecular Weight : 122.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-fluoropropanedioic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75453
Availability details for
Quantity
  • Availability
  • Add to Cart