ADVH49B82030 AChemBlock

1H-benzo[d]imidazole-4-carboxylic acid

  • Cas Number : 46006-36-4
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1h-benzo-d-imidazole-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71325
Availability details for
Quantity
  • Availability
  • Add to Cart