ADVH49B82038 AChemBlock

3-Chloro-4-methylphenylboronic acid pinacol ester

  • Cas Number : 445303-10-6
  • Molecular Weight : 270.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/3-chloro-4-methylphenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70724
Availability details for
Quantity
  • Availability
  • Add to Cart