ADVH49B8203C AChemBlock

2-(4-Bromophenoxy)-1-{4-[(4-fluorophenyl)sulfonyl]piperazin-1-yl}ethanone

  • Cas Number : 439939-70-5
  • Molecular Weight : 457.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/2-4-bromophenoxy-1-4-4-fluorophenyl-sulfonyl-piperazin-1-yl-ethanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86763
Availability details for
Quantity
  • Availability
  • Add to Cart