ADVH49B8203E AChemBlock

Lasmiditan hemisuccinate

  • Cas Number : 439239-92-6
  • Molecular Weight : 872.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/lasmiditan-hemisuccinate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72361
Availability details for
Quantity
  • Availability
  • Add to Cart