ADVH49B82040 AChemBlock

BCI-121

  • Cas Number : 432529-82-3
  • Molecular Weight : 340.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/bci-121.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86764
Availability details for
Quantity
  • Availability
  • Add to Cart