ADVH49B82045 AChemBlock

2,4-dichloro-6-methylpyridine

  • Cas Number : 42779-56-6
  • Molecular Weight : 162.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-dichloro-6-methylpyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71339
Availability details for
Quantity
  • Availability
  • Add to Cart