ADVH49B8204A AChemBlock

tert-butyl 4-(3-oxobutanoyl)piperidine-1-carboxylate

  • Cas Number : 419571-68-9
  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-3-oxobutanoyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75608
Availability details for
Quantity
  • Availability
  • Add to Cart