ADVH49B8204B AChemBlock

tetralin-5-carbaldehyde

  • Cas Number : 41828-13-1
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tetralin-5-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75616
Availability details for
Quantity
  • Availability
  • Add to Cart