ADVH49B82056 AChemBlock

methyl 3-methylpyrazine-2-carboxylate

  • Cas Number : 41110-29-6
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-methylpyrazine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75654
Availability details for
Quantity
  • Availability
  • Add to Cart