ADVH49B82058 AChemBlock

5-Chloro-1H-imidazo[4,5-b]pyridin-2(3H)-one

  • Cas Number : 40851-98-7
  • Molecular Weight : 169.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-1h-imidazo-4-5-b-pyridin-2-3h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86491
Availability details for
Quantity
  • Availability
  • Add to Cart