ADVH49B82059 AChemBlock

1-tert-butoxycarbonyl-4-methylene-piperidine-2-carboxylic acid

  • Cas Number : 407627-05-8
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butoxycarbonyl-4-methylene-piperidine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75675
Availability details for
Quantity
  • Availability
  • Add to Cart