ADVH49B8205D AChemBlock

3,5,6-trichloropicolinic acid

  • Cas Number : 40360-44-9
  • Molecular Weight : 226.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-5-6-trichloropicolinic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86506
Availability details for
Quantity
  • Availability
  • Add to Cart