ADVH49B82069 AChemBlock

8-fluoroquinoline

  • Cas Number : 394-68-3
  • Molecular Weight : 147.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-fluoroquinoline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71367
Availability details for
Quantity
  • Availability
  • Add to Cart