ADVH49B8206B AChemBlock

1-indan-4-ylethanone

  • Cas Number : 38997-97-6
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-indan-4-ylethanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75789
Availability details for
Quantity
  • Availability
  • Add to Cart