ADVH49B8206F AChemBlock

N,N-Dimethyl-2-piperidinecarboxamide hydrochloride

  • Cas Number : 383425-11-4
  • Molecular Weight : 192.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/n-n-dimethyl-2-piperidinecarboxamide-hydrochloride-293647.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70337
Availability details for
Quantity
  • Availability
  • Add to Cart