ADVH49B82074 AChemBlock

1-(2,2,2-trifluoroethyl)-3H-imidazo[4,5-c]pyridin-2-one

  • Cas Number : 380605-32-3
  • Molecular Weight : 217.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-2-2-2-trifluoroethyl-3h-imidazo-4-5-c-pyridin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75835
Availability details for
Quantity
  • Availability
  • Add to Cart