ADVH49B82075 AChemBlock

2-Methylsulfonylaminophenylboronic acid, pinacol ester

  • Cas Number : 380430-60-4
  • Molecular Weight : 315.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-methylsulfonylaminophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70489
Availability details for
Quantity
  • Availability
  • Add to Cart