ADVH49B82077 AChemBlock

ethyl 1H-pyrrole-3-carboxylate

  • Cas Number : 37964-17-3
  • Molecular Weight : 139.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1h-pyrrole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75844
Availability details for
Quantity
  • Availability
  • Add to Cart