ADVH49B82079 AChemBlock

ethyl 6-bromoimidazo[1,2-a]pyridine-3-carboxylate

  • Cas Number : 372198-69-1
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-6-bromoimidazo-1-2-a-pyridine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75874
Availability details for
Quantity
  • Availability
  • Add to Cart