ADVH49B8207A AChemBlock

5-bromopyrimidine-2-carboxylic acid

  • Cas Number : 37131-87-6
  • Molecular Weight : 203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-bromopyrimidine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75879
Availability details for
Quantity
  • Availability
  • Add to Cart