ADVH49B8207D AChemBlock

1-((isocyano(phenyl)methyl)sulfonyl)-4-methylbenzene

  • Cas Number : 36635-66-2
  • Molecular Weight : 271.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-isocyano-phenyl-methyl-sulfonyl-4-methylbenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86569
Availability details for
Quantity
  • Availability
  • Add to Cart