ADVH49B8207F AChemBlock

tert-butyl 5-formyl-2,2-dimethyl-1,3-dioxan-5-ylcarbamate

  • Cas Number : 364631-73-2
  • Molecular Weight : 259.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-formyl-2-2-dimethyl-1-3-dioxan-5-ylcarbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86745
Availability details for
Quantity
  • Availability
  • Add to Cart