ADVH49B82081 AChemBlock

1-[4-(cyclopentylamino)-2-(methylsulfanyl)pyrimidin-5-yl]ethan-1-one

  • Cas Number : 362656-11-9
  • Molecular Weight : 251.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-4-cyclopentylamino-2-methylsulfanyl-pyrimidin-5-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75911
Availability details for
Quantity
  • Availability
  • Add to Cart