ADVH49B82083 AChemBlock

6-methyl-1,2,3,4-tetrahydroquinolin-4-one

  • Cas Number : 36054-00-9
  • Molecular Weight : 161.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methyl-1-2-3-4-tetrahydroquinolin-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75925
Availability details for
Quantity
  • Availability
  • Add to Cart