ADVH49B82088 AChemBlock

2-chloro-4-(pyrrolidin-1-yl)pyrimidine

  • Cas Number : 35691-20-4
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-4-pyrrolidin-1-yl-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71401
Availability details for
Quantity
  • Availability
  • Add to Cart