ADVH49B8208E AChemBlock

2-bromo-6-chlorobenzo[d]thiazole

  • Cas Number : 3507-17-3
  • Molecular Weight : 248.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-6-chlorobenzo-d-thiazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86552
Availability details for
Quantity
  • Availability
  • Add to Cart