ADVH49B82091 AChemBlock

4-(trifluoromethyl)-1,3-thiazol-2-amine

  • Cas Number : 349-49-5
  • Molecular Weight : 168.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-trifluoromethyl-1-3-thiazol-2-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75983
Availability details for
Quantity
  • Availability
  • Add to Cart