ADVH49B82093 AChemBlock

7-chloro-2,3-dihydro-1H-inden-1-one

  • Cas Number : 34911-25-6
  • Molecular Weight : 166.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-chloro-2-3-dihydro-1h-inden-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75986
Availability details for
Quantity
  • Availability
  • Add to Cart