ADVH49B82095 AChemBlock

2-ethyl-1H-indole

  • Cas Number : 3484-18-2
  • Molecular Weight : 145.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-ethyl-1h-indole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S75988
Availability details for
Quantity
  • Availability
  • Add to Cart