ADVH49B8209A AChemBlock

PJ34

  • Cas Number : 344458-19-1
  • Molecular Weight : 295.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/pj34.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86672
Availability details for
Quantity
  • Availability
  • Add to Cart