ADVH49B8209C AChemBlock

1-(6-fluoro-1H-indol-3-yl)-N,N-dimethylmethanamine

  • Cas Number : 343-93-1
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-6-fluoro-1h-indol-3-yl-n-n-dimethylmethanamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86387
Availability details for
Quantity
  • Availability
  • Add to Cart