ADVH49B8209E AChemBlock

2-(1-bicyclo[2.2.2]octanyl)acetonitrile

  • Cas Number : 34341-70-3
  • Molecular Weight : 149.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-1-bicyclo-2-2-2-octanyl-acetonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76023
Availability details for
Quantity
  • Availability
  • Add to Cart