ADVH49B820A1 AChemBlock

tert-butyl (2R)-2-ethenylpyrrolidine-1-carboxylate

  • Cas Number : 340129-94-4
  • Molecular Weight : 197.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2r-2-ethenylpyrrolidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76036
Availability details for
Quantity
  • Availability
  • Add to Cart