ADVH49B820A8 AChemBlock

2-chloro-5-methyl-4-(trifluoromethyl)pyridine

  • Cas Number : 326894-70-6
  • Molecular Weight : 195.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-5-methyl-4-trifluoromethyl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76101
Availability details for
Quantity
  • Availability
  • Add to Cart