ADVH49B820A9 AChemBlock

6-chloro-2,3-dihydro-1-benzofuran-3-one

  • Cas Number : 3260-78-4
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-2-3-dihydro-1-benzofuran-3-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76102
Availability details for
Quantity
  • Availability
  • Add to Cart