ADVH49B820AD AChemBlock

benzofuran-3-ylboronic acid

  • Cas Number : 317830-83-4
  • Molecular Weight : 161.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/benzofuran-3-ylboronic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76138
Availability details for
Quantity
  • Availability
  • Add to Cart