ADVH49B820B0 AChemBlock

2-Thioxo-3-[3-(trifluoromethyl)phenyl]-4-thiazolidinon

  • Cas Number : 315-08-2
  • Molecular Weight : 277.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-thioxo-3-3-trifluoromethyl-phenyl-4-thiazolidinon.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86756
Availability details for
Quantity
  • Availability
  • Add to Cart