ADVH49B820B3 AChemBlock

(1R)-1-pyrimidin-2-ylethanol

  • Cas Number : 31415-75-5
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1r-1-pyrimidin-2-ylethanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76158
Availability details for
Quantity
  • Availability
  • Add to Cart