ADVH49B820B5 AChemBlock

ethyl 2-(tert-butoxycarbonylamino)-2-(4-pyridyl)acetate

  • Cas Number : 313490-99-2
  • Molecular Weight : 280.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-tert-butoxycarbonylamino-2-4-pyridyl-acetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S76167
Availability details for
Quantity
  • Availability
  • Add to Cart