ADVH49B820BD AChemBlock

2-(4-oxo-3,4-dihydroquinazolin-2-yl)acetonitrile

  • Cas Number : 30750-23-3
  • Molecular Weight : 185.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-oxo-3-4-dihydroquinazolin-2-yl-acetonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86705
Availability details for
Quantity
  • Availability
  • Add to Cart