ADVH49B820BE AChemBlock

3-bromo-4-fluorobiphenyl

  • Cas Number : 306935-88-6
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-fluorobiphenyl.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71453
Availability details for
Quantity
  • Availability
  • Add to Cart