ADVH49B820BF AChemBlock

3-Methanesulfonylaminophenylboronic acid, pinacol ester

  • Cas Number : 305448-92-4
  • Molecular Weight : 315.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/3-methanesulfonylaminophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70490
Availability details for
Quantity
  • Availability
  • Add to Cart